missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H3N2 Hemagglutinin Antibody (1B7), Alexa Fluor™ 350, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Influenza A H3N2 Hemagglutinin Monoclonal antibody specifically detects Influenza A H3N2 Hemagglutinin in Influenza A virus H3N2 samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Influenza A H3N2 Hemagglutinin |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1B7 |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Recombinant Influenza A Virus Hemagglutinin (17-345aa) purified from Baculovirus |
| Purification Method | Protein G purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?