missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1pdm09 NP (A/California/7/2009) Recombinant Rabbit Monoclonal Antibody (HL1098), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA546926
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The H1N1pdm09 virus completely replaced the previously circulating seasonal H1N1 IAV and continued to circulate during subsequent influenza seasons|The M1 protein of Influenza A virus H1N1pdm09 has an "e;avian signature"e; and lacks the extra-epitopic amino acid residues seen in previous human Influenza A viruses [PMID: 29659927]Specifications
Influenza A H1N1pdm09 NP (A/California/7/2009) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Western Blot | |
HL1098 | |
Unconjugated | |
H1N1 NP; Influenza A; Influenza H1N1 | |
Recombinant protein encompassing a sequence within the C-terminus region of Influenza A virus H1N1pdm09 Nucleoprotein (A/California/7/2009). The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |