missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Nucleoprotein Antibody (GT1236) - (A/WSN/1933), Novus Biologicals™
Shop All Bio Techne ProductsDescription
Influenza A H1N1 Nucleoprotein Monoclonal antibody specifically detects Influenza A H1N1 Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
Specifications
Specifications
| Antigen | Influenza A H1N1 Nucleoprotein |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA |
| Classification | Monoclonal |
| Clone | GT1236 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Recombinant protein of Influenza A H1N1 nucleoprotein (A/WSN/1933). The exact sequence is proprietary. |
| Purification Method | Protein G purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?