missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H1N1 Nucleoprotein Antibody (2F205), DyLight 594, Novus Biologicals™

Product Code. 18417327 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417327 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417327 Supplier Novus Biologicals Supplier No. NBP224871DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Influenza A H1N1 Nucleoprotein Monoclonal antibody specifically detects Influenza A H1N1 Nucleoprotein in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H1N1 Nucleoprotein
Applications ELISA
Classification Monoclonal
Clone 2F205
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Recombinant influenza A H1N1 protein was used as the immunogen
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.