missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 NS1 Antibody (GT1943) - (A/WSN/1933), Novus Biologicals™
Description
Influenza A H1N1 NS1 Monoclonal antibody specifically detects Influenza A H1N1 NS1 in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Influenza A H1N1 NS1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | GT1943 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | Influenza NS1 |
| Host Species | Mouse |
| Immunogen | Recombinant protein of Influenza A Virus NS1 (nonstructural protein) (A/WSN/1933(H1N1). The exact sequence is proprietary. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?