missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 Hemagglutinin Antibody (IVC102) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1483
This item is not returnable.
View return policy
Description
Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza A Virus H1N1, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Hemagglutination Assay.
Specifications
| Influenza A H1N1 Hemagglutinin | |
| Monoclonal | |
| 4.2 mg/mL | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Haemagglutination | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Influenza A H1N1. Reacts with hemagglutinin of H1N1 serotype. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Hemagglutination Assay | |
| IVC102 | |
| Unconjugated | |
| HA, Hemagglutinin | |
| Purified influenza virus type A strain H1N1. | |
| 0.5 mg | |
| Virology Bacteria and Parasites | |
| 956529 | |
| Influenza virus, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction