missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H17N10 Hemagglutinin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12749-100ul
This item is not returnable.
View return policy
Description
Influenza A H17N10 Hemagglutinin Polyclonal antibody specifically detects Influenza A H17N10 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Influenza A H17N10 Hemagglutinin | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Influenza A H17N10 Hemagglutinin (Accession#: AFC35418.1; Met1-Tyr527). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction