missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H15N8 Hemagglutinin Rabbit anti-Virus, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Influenza A H15N8 Hemagglutinin Polyclonal antibody specifically detects Influenza A H15N8 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Influenza A H15N8 Hemagglutinin |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 |
| Formulation | 0.2μm filtered solution in PBS without Preservative |
| Host Species | Rabbit |
| Immunogen | Purified, recombinant Influenza A H15N8 Hemagglutinin (ABB88132.1; Met1-Val534) |
| Purification Method | Protein A purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?