Learn More
ACROBiosystems Influenza A [A/Hong Kong/483/97 (H5N1)] Hemagglutinin (HA) Protein, His Tag
Description
Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Hemagglutinin also plays a major role in the determination of host range restriction and virulence. As a class I viral fusion protein, hemagglutinin is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane.
Specifications
Specifications
| Molecular Weight (g/mol) | 60.1 kDa |
| Quantity | 1 mg |
| Source | Influenza A [A/Hong Kong/483/97 (H5N1)] HA, His Tag (HA1-V5229) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Gln 531 (Accession # AAC32099.1). |
| Immunogen | Asp 17 - Gln 531 |
| Storage Requirements | -20°C |
| Gene Alias | HA,Hemagglutinin |
| Common Name | Hemagglutinin (HA) |
| Conjugate | Unconjugated |
| Species | Influenza |
| Protein Tag | His Tag |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.