Learn More
Invitrogen™ Infliximab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558408
 View more versions of this product
Product Code. 30284326
  • £1002.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Infliximab with the brand name of Remicade, is a chimeric monoclonal IgG1 antibody. It binds to TNF-alpha, which is a key proinflammatory cytokine involved in chronic inflammatory diseases. Infliximab functions as TNF-alpha blocker to disrupt the interaction of TNF-alpha with its receptors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Infliximab Chimeric
Recombinant Monoclonal
Unconjugated
Remicade; TA-650
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized