missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Infliximab Chimeric Recombinant Human Monoclonal Antibody (cA2 (Infliximab))

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547798
This item is not returnable.
View return policy
Description
Specificity: Binds to soluble and transmembrane forms of TNF-alpha and neutralizes its biological activity. Does not bind to TNF-beta.
Infliximab with the brand name of Remicade, is a chimeric monoclonal IgG1 antibody. It binds to TNF-alpha, which is a key proinflammatory cytokine involved in chronic inflammatory diseases. Infliximab functions as TNF-alpha blocker to disrupt the interaction of TNF-alpha with its receptors.
Specifications
| Infliximab Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P08183 | |
| Human | |
| Protein A | |
| RUO | |
| 5243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Functional Assay, Western Blot | |
| cA2 (Infliximab) | |
| Unconjugated | |
| ABCB1 | |
| Remicade; TA-650 | |
| Purified recombinant human TNF (rTNF). | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction