missing translation for 'onlineSavingsMsg'
Learn More

INF2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18409356 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409356 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409356 Supplier Novus Biologicals Supplier No. NBP281836AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

INF2 Polyclonal antibody specifically detects INF2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen INF2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias C14orf151, C14orf173DKFZp762A0214, chromosome 14 open reading frame 151, chromosome 14 open reading frame 173, FLJ22056, FSGS5, HBEAG-binding protein 2 binding protein C, HBEBP2-binding protein C, inverted formin, FH2 and WH2 domain containing, inverted formin-2, MGC13251, pp9484
Host Species Rabbit
Immunogen INF2 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human INF2. Amino Acid Squence: KEKLGPQDSDPTEANLES
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 64423
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.