Learn More
BINDER™ Inert Gas Connection with Gas Inlet and Outlet
Product Code. 17844431 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Series FP 115 E3 Avantgarde
BINDER Series FP 260 Avantgarde
BINDER Series FP Avantgarde
Unit Size:
Each
Product Code. 17844431
Brand: BINDER™ 80122195
Only null left
Add to basket
Add to basket
The inert gas connection includes additional measures for the greatest possible impermeability to gas.
  • Accessory for Series FP Avantgarde Line Drying and Heating Chambers
Specifications
For Use With (Equipment) BINDER Series FP Avantgarde
Type Connection Kit
Includes Gas Inlet and Outlet
Diameter (English) 0.39 in.
Diameter (Metric) 10 mm
Voltage 120/230 V