missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Inert Gas Connection

Product Code. 16404389 Shop All BINDER Products
Click to view available options
Diameter (Metric):
10 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16404389

Brand: BINDER™ 80121693

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: FD 260

  • With gas inlet and outlet
  • Not in conjunction with access port or interior lighting option

Specifications

Description Inert gas connection - With gas inlet and outlet, Ø 10 mm, incl. additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option
For Use With (Equipment) BINDER Chambers FD 260
Material Steel
Type Gas Connection
Diameter (English) 0.4 in.
Diameter (Metric) 10 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.