missing translation for 'onlineSavingsMsg'
Learn More

Manutan Industrial locker with feet, cylinder lock, grey, one tier, three nest

Brand:  Manutan 1098O054

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12834558

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Fitted with 150mm high feet for improved ventilation and easy cleaning. One tier lockers with hat shelf, hanging rail and three coat hooks. Two tier lockers with hanging rail and three coat hooks. Industrial quality with powder coated finish. Doors with sound reducers, ventilation slots and label holder.
  • Three lockers supplied
  • One tier
  • Cylinder lock
  • Grey door
  • Dimensions (H x W x D): 1850mm x 930mm x 500mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers