Learn More
Sherwood Scientific™ Industrial Flame Photometer
Product Code. 12981801
Click to view available options
Model:
M420
Unit Size:
Each
Product Code. 12981801
Brand: Sherwood Scientific™ 47542100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Industrial single/dual element displayed 240V 50Hz Na with K, Na with Li 390mm x 345mm x 510mm Sherwood Scientific
Specifications
Model M420