missing translation for 'onlineSavingsMsg'
Learn More

Indoxyl-beta-D-glucoside, 98%

Product Code. 15542353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15542353 250 mg 250mg
15532353 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15542353 Supplier Thermo Scientific Acros Supplier No. 458882500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 487-60-5 | C14H17NO6 | 295.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-60-5
Molecular Formula C14H17NO6
Molecular Weight (g/mol) 295.29
MDL Number MFCD00047169
InChI Key XVARCVCWNFACQC-RKQHYHRCSA-N
Synonym indican, 3-indolyl-b-d-glucopyranoside, uroxanthin, 3-indoxyl-beta-d-glucopyranoside, indoxyl, a-d-glucoside, unii-n187wk1y1j, indikan, indican, plant, indican glucoside, 3-indolyl b-d-glucopyranoside
PubChem CID 441564
ChEBI CHEBI:16700
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 57.0°C to 58.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol, alcohol, acetone, slightly soluble in ether, benzene, chloroform,, ethyl acetate, carbon disulfide
Formula Weight 295.29
Percent Purity 98%
Chemical Name or Material Indoxyl-β-D-glucoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.