missing translation for 'onlineSavingsMsg'
Learn More

Indoxyl-Beta-D-glucoside, TRC

Product Code. 30400650
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400650 100 mg 100mg
30392912 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400650 Supplier Toronto Research Chemicals Supplier No. TRCI654110100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 487-60-5
Molecular Formula C14H17NO6
Molecular Weight (g/mol) 295.29
Synonym 1H-Indol-3-yl beta-D-glucopyranoside, 3-(Glucosyloxy)indole, Indican (Glucoside), NSC 87517, Uroxanthin
SMILES OC[C@H]1O[C@@H](Oc2c[nH]c3ccccc23)[C@H](O)[C@@H](O)[C@@H]1O

Specifikationer

Quantity 100 mg
InChI Formula InChI=1S/C14H17NO6/c16-6-10-11(17)12(18)13(19)14(21-10)20-9-5-15-8-4-2-1-3-7(8)9/h1-5,10-19H,6H2/t10-,11-,12+,13-,14-/m1/s1
Chemical Name or Material Indoxyl-beta-D-glucoside
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.