missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indoximod, MedChemExpress
Description
Indoximod (1-Methyl-D-tryptophan) is an orally active indoleamine 2,3-dioxygenase (IDO) pathway inhibitor. Indoximod acts as a Trp mimetic in regulating mTOR. Indoximod is an immunometabolic adjuvant used for the research of cancer.
Specifications
Specifications
| Chemical Name or Material | Indoximod |
| CAS | 110117-83-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H14N2O2 |
| Quantity | 250 mg |
| Synonym | 1-Methyl-D-tryptophan NLG-8189 |
| Solubility Information | H2O : 5 mg/mL (22.91 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 0.55 mg/mL (2.52 mM; Need ultrasonic and warming) |
| SMILES | N[C@H](CC1=CN(C)C2=CC=CC=C12)C(O)=O |
| Molecular Weight (g/mol) | 218.25 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?