missing translation for 'onlineSavingsMsg'
Learn More

Indoprofen, MedChemExpress

Product Code. 16498606
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498606 100 mg 100mg
16458668 10 mg 10mg
16451098 5 mg 5mg
16434547 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498606 Supplier Medchem Express Supplier No. HYB1104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Indoprofen is a non-steroidal anti-inflammatory drug, provide insight into treatments for spinal muscular atrophies.

Indoprofen is a non-steroidal anti-inflammatory drug, provide insight into treatments for spinal muscular atrophies.

Specifications

Chemical Name or Material Indoprofen
CAS 31842-01-0
Color White
Purity Grade Notes Research
Molecular Formula C17H15NO3
Quantity 100 mg
Synonym (±)-Indoprofe
Solubility Information DMSO : 100 mg/mL (355.48 mM; Need ultrasonic)
SMILES O=C(O)C(C)C1=CC=C(N(CC2=C3C=CC=C2)C3=O)C=C1
Molecular Weight (g/mol) 281.31
Formula Weight 281.31
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.