missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin, TRC

Product Code. 18031520
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18031520 50 mg 50mg
18093510 1 g 1g
18042920 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18031520 Supplier Toronto Research Chemicals Supplier No. TRCI64100050MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-86-1
Molecular Formula C19 H16 Cl N O4
Molecular Weight (g/mol) 357.79
Synonym 1H-Indole-3-acetic acid, 1-(4-chlorobenzoyl)-5-methoxy-2-methyl- (9CI, ACI), 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid (ACI), Indole-3-acetic acid, 1-(p-chlorobenzoyl)-5-methoxy-2-methyl- (8CI), 1-(4-Chlorobenzoyl)-2-methyl-5-methoxyindole-3-acetic acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid, 1-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetic acid, 1-(p-Chlorobenzoyl)-2-methyl-5-methoxyindole-3-acetic acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methyl-3-indolylacetic acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid, 2-[1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid, 2-[1-(4-Chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid, 2-[1-[(4-Chlorophenyl)carbonyl]-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid, [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid, Aconip, Amuno, Argun, Artracin, Artrinovo, Artrivia, Bonidin, Bonidon Gel, Catlep, Chibro-Amuno, Chrono-Indicid, Chrono-Indocid 75, Confortid, Dolcidium, Dolcidium PL, Dolovin, Durametacin, Elmetacin, Endol, Flam, Globa, Idomethine, Imbrilon, IMC, Inacid, Indacin, Indo-Phlogont, Indo-Rectolmin, Indo-Tablinen, Indocid, Indocid (pharmaceutical), Indocin, Indocin IV, Indocine, Indocollyre, Indomecol, Indomed, Indomee, Indometacin, Indometacine, Indomethacin, Indomethacine, Indomethine, Indomod, Indonol, Indoptic, Indoptol, Indorektal, IndoRich, Indoxen, Indren, Inflazon, Infrocin, Innamit, Inteban, Inteban SP, Lausit, MeSH ID: D007213, Metacen, Metartril, Methazine, Metindol, Mezolin, Microcid, Mikametan, Mobilan, N-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetic acid, NSC 77541, Reumacide, Rheumacin LA, Sadoreum, Tannex, Vital Vitacid, Vonum, α-[1-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolyl]acetic acid
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
SMILES COc1ccc2c(c1)c(CC(=O)O)c(C)n2C(=O)c3ccc(Cl)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)
Formula Weight 357.0768
Percent Purity >95%
Chemical Name or Material Indomethacin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.