missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin Methyl Ester, TRC

Product Code. 30404735
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404735 5 mg 5mg
30392560 50 mg -
30401119 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30404735 Supplier Toronto Research Chemicals Supplier No. TRCI6410255MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1601-18-9
Molecular Formula C20 H18 Cl N O4
Molecular Weight (g/mol) 371.81
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid methyl ester, Indole-3-acetic acid, 1-(p-chlorobenzoyl)-5-methoxy-2-methyl-, methyl ester (7CI,8CI), Indomethacin methyl ester, L 588983, Methyl 1-(4-chlorobenzoyl)-5-methoxy-2-methylindole-3-acetate, Methyl N-(p-chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetate
IUPAC Name methyl 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
SMILES COC(=O)Cc1c(C)n(C(=O)c2ccc(Cl)cc2)c3ccc(OC)cc13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H18ClNO4/c1-12-16(11-19(23)26-3)17-10-15(25-2)8-9-18(17)22(12)20(24)13-4-6-14(21)7-5-13/h4-10H,11H2,1-3H3
Formula Weight 371.0924
Chemical Name or Material Indomethacin Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.