missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin Isopropyl Ester, Mikromol™

Product Code. 30050570
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050570 25 mg 25mg
30051114 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050570 Supplier Mikromol Supplier No. MM0584.110025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Indomethacin Isopropyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indomethacin Isopropyl Ester
CAS 72616-25-2
Analyte or Component Names Indomethacin Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C22 H22 Cl N O4
Certifications/Compliance ISO 17025
Synonym Indomethacin Isopropyl Ester, 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid 1-methylethyl ester
InChI Formula InChI=1S/C22H22ClNO4/c1-13(2)28-21(25)12-18-14(3)24(20-10-9-17(27-4)11-19(18)20)22(26)15-5-7-16(23)8-6-15/h5-11,13H,12H2,1-4H3
SMILES COc1ccc2c(c1)c(CC(=O)OC(C)C)c(C)n2C(=O)c3ccc(Cl)cc3
IUPAC Name propan-2-yl 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
Molecular Weight (g/mol) 399.87
Quantity 25 mg
Formula Weight 399.1237 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.