missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin Diamide, TRC

Product Code. 30393744
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393744 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393744 Supplier Toronto Research Chemicals Supplier No. TRCI641020100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indomethacin Diamide
CAS 402849-25-6
Molecular Formula C33H27Cl2N3O5
Quantity 100 mg
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-Indole-3-acetic Acid 2-(4-chlorobenzoyl)-2-(4-methoxyphenyl)hydrazide, Indomethacin Impurity J
InChI Formula InChI=1S/C33H27Cl2N3O5/c1-20-28(29-18-27(43-3)16-17-30(29)37(20)32(40)21-4-8-23(34)9-5-21)19-31(39)36-38(25-12-14-26(42-2)15-13-25)33(41)22-6-10-24(35)11-7-22/h4-18H,19H2,1-3H3,(H,36,39)
SMILES O=C(C1=CC=C(Cl)C=C1)N(NC(CC2=C(C)N(C(C3=CC=C(Cl)C=C3)=O)C4=C2C=C(OC)C=C4)=O)C5=CC=C(OC)C=C5
IUPAC Name 4-chloro-N'-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]-N-(4-methoxyphenyl)benzohydrazide
Molecular Weight (g/mol) 616.49
Formula Weight 615.1328
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.