missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin Acyl-Beta-D-glucopyranuronic Acid Phenyl Ester, TRC

Product Code. 30393743
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393743 25 mg 25mg
30388931 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393743 Supplier Toronto Research Chemicals Supplier No. TRCI64101525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indomethacin Acyl-beta-D-glucopyranuronic Acid Phenyl Ester
Analyte or Component Names Indomethacin Acyl-beta-D-glucopyranuronic Acid Phenyl Ester
Shipping Condition Room Temperature
Molecular Formula C32H30ClNO10
InChI Formula InChI=1S/C32H30ClNO10/c1-17-22(23-14-21(41-2)12-13-24(23)34(17)30(39)19-8-10-20(33)11-9-19)15-25(35)43-32-28(38)26(36)27(37)29(44-32)31(40)42-16-18-6-4-3-5-7-18/h3-14,26-29,32,36-38H,15-16H2,1-2H3/t26-,27-,28+,29-,32+/m1/s1
Solution Type Neat
SMILES O=C(O[C@H]([C@H]1O)O[C@@H](C(OCC2=CC=CC=C2)=O)[C@@H]([C@H]1O)O)CC3=C(C)N(C(C4=CC=C(Cl)C=C4)=O)C5=C3C=C(OC)C=C5
IUPAC Name benzyl (2R,3R,4R,5S,6R)-6-(2-(1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetoxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 624.03
Quantity 25 mg
Formula Weight 623.16
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.