missing translation for 'onlineSavingsMsg'
Learn More

Indomethacin, 98%

Product Code. 11406390
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406390 25 g 25g
11496380 5 g 5g
11416390 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406390 Supplier Thermo Scientific Alfa Aesar Supplier No. A19910.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53-86-1 | C19H16ClNO4 | 357.79 g/mol

It is a non-steroidal anti-inflammatory drug, it is used in the treatment of musculoskeletal and joint disorders including osteoarthritis, rheumatoid arthritis, gout, bursitis and tendinitis. It finds its application as gout suppressant, non-steroidal anti-inflammatory drug and analgesic. It also plays many biological roles. Additionally, Indomethacin displays adipogenic actions via binding and activation of PPARγ (peroxisome proliferator-activated receptor γ), a ligand-activated transcription factor known to play an important role in adipogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in acetone (40 mg/mL - clear, yellow solution), ethanol (20 mg/mL), ether, castor oil; Soluble in chloroform (50 mg/mL - clear, yellow, extremely viscous solution); decomposed by strong alkali but stable in neutral or slightly acidic media; insoluble in water.

Notes
Store away from oxidizing agents, strong bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-86-1
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
MDL Number MFCD00057095
InChI Key CGIGDMFJXJATDK-UHFFFAOYSA-N
Synonym indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
PubChem CID 3715
ChEBI CHEBI:49662
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O

Specifications

Melting Point 158°C to 161°C
Assay Percent Range 0.98
Quantity 25 g
UN Number UN2811
Beilstein 497341
Sensitivity Light sensitive
Merck Index 14,4968
Solubility Information Soluble in acetone (40mg/mL- clear,yellow solution),ethanol (20mg/mL),ether,castor oil; Soluble in chloroform (50mg/mL- clear,yellow,extremely viscous solution); decomposed by strong alkali but stable in neutral or slightly acidic media; insoluble in water.
Formula Weight 357.8
Percent Purity 98%
Chemical Name or Material Indomethacin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.