missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Indoline-2-carboxylic acid, 95%

Product Code. 11447136
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447136 5 g 5g
11457136 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447136 Supplier Thermo Scientific Alfa Aesar Supplier No. B25450.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78348-24-0 | C9H8NO2 | 162.17 g/mol

(±)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78348-24-0
Molecular Formula C9H8NO2
Molecular Weight (g/mol) 162.17
MDL Number MFCD00010635
InChI Key QNRXNRGSOJZINA-QMMMGPOBSA-M
Synonym indoline-2-carboxylic acid, +/--indoline-2-carboxylic acid, dl-indoline-2-carboxylic acid, 1h-indole-2-carboxylicacid, 2,3-dihydro, 2,3-dihydroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro, 2-indolinecarboxylic acid, indoline-2-carboxylicacid, 1-indoline-2-carboxylic acid, pubchem3035
PubChem CID 86074
IUPAC Name 2,3-dihydro-1H-indole-2-carboxylic acid
SMILES [O-]C(=O)[C@@H]1CC2=CC=CC=C2N1

Specifications

Melting Point ∼168°C (decomposition)
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 163.18
Percent Purity 95%
Chemical Name or Material (+/-)-Indoline-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.