missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indoleamine 2,3-dioxygenase/IDO Antibody (BLR040F), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-76408
This item is not returnable.
View return policy
Description
Indoleamine 2,3-dioxygenase/IDO Monoclonal specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Indoleamine 2,3-dioxygenase/IDO | |
| Monoclonal | |
| Unconjugated | |
| Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA with 0.09% Sodium Azide | |
| IDO1 | |
| Recombinant protein representing Human IDO1 | |
| 100 μL | |
| 3620.0 | |
| Store at 2-8°C/ 1 year from date of receipt | |
| IgG |
| Western Blot, Immunoprecipitation | |
| BLR040F | |
| Western Blot 1:1000, Immunoprecipitation 15 μL/1mg lysate | |
| EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase | |
| Rabbit | |
| Antigen Affinity-purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction