missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indoleamine 2,3-dioxygenase/IDO Antibody (998736) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB60302JF585
This item is not returnable.
View return policy
Description
Indoleamine 2,3-dioxygenase/IDO Monoclonal antibody specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Indoleamine 2,3-dioxygenase/IDO | |
| Monoclonal | |
| Janelia Fluor 585 | |
| EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase | |
| E. coli-derived human Indoleamine 2,3-dioxygenase/IDO, Ala2-Gly403, Accession # P14902 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 998736 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Myeloid derived Suppressor Cell | |
| 3620 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction