Learn More
Indole, 99+%, Thermo Scientific Chemicals
£16.40 - £640.00
Chemical Identifiers
CAS | 120-72-9 |
---|---|
MDL Number | MFCD00005607 |
InChI Key | SIKJAQJRHWYJAI-UHFFFAOYSA-N |
Synonym | indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german |
PubChem CID | 798 |
ChEBI | CHEBI:16881 |
IUPAC Name | 1H-indole |
SMILES | C1=CC=C2C(=C1)C=CN2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10162680
|
Thermo Scientific Acros
122150100 |
10 g | Glass Bottle |
£16.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10295300
|
Thermo Scientific Acros
122150250 |
25 g | Glass bottle |
£20.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10205310
|
Thermo Scientific Acros
122151000 |
100 g | Glass Bottle |
£48.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041240
|
Thermo Scientific Acros
122155000 |
500 g | Glass Bottle |
£165.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13488328
|
Thermo Scientific Acros
122150025 |
2.5 kg | Plastic bottle |
£640.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
120-72-9 | |
SIKJAQJRHWYJAI-UHFFFAOYSA-N | |
798 | |
1H-indole |
MFCD00005607 | |
indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german | |
CHEBI:16881 | |
C1=CC=C2C(=C1)C=CN2 |
Specifications
120-72-9 | |
Pink to White | |
253.0°C to 254.0°C | |
Authentic | |
C8H7N | |
20, 304 | |
1.22 | |
Solubility in water: 2.80g/L (25°C). Other solubilities: soluble in hot water,soluble in hot alcohol,ether and benzene,0.1g/mL meoh | |
C1=CC=C2C(=C1)C=CN2 | |
117.15 | |
CHEBI:16881 | |
99+% | |
Indole, 98+% |
51.0°C to 53.0°C | |
1.2200g/mL | |
136°C | |
99+% | |
MFCD00005607 | |
15, 5003 | |
indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german | |
SIKJAQJRHWYJAI-UHFFFAOYSA-N | |
1H-indole | |
798 | |
117.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-420-7
missing translation for 'rtecsNumber' : NL2450000
missing translation for 'tsca' : TSCA
- Indole
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only