missing translation for 'onlineSavingsMsg'
Learn More

Indole-7-carboxaldehyde, 98%

Product Code. 11318466
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318466 250 mg 250mg
11308466 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318466 Supplier Thermo Scientific Alfa Aesar Supplier No. L17506.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1074-88-0 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-88-0
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD01318152
InChI Key XQVZDADGTFJAFM-UHFFFAOYSA-N
Synonym indole-7-carboxaldehyde, 7-formylindole, indole-7-carbaldehyde, 7-formyl-1h-indole, 1h-indole-7-carboxaldehyde, indole-7-carboxaldehyde 7-formylindole, 7-formyl indole, 7-indolcarboxaldehyde, pubchem7270, 7-indolecarboxaldehyde
PubChem CID 2734629
IUPAC Name 1H-indole-7-carbaldehyde
SMILES O=CC1=C2NC=CC2=CC=C1

Specifications

Melting Point 86°C to 89°C
Quantity 250 mg
Beilstein 1524960
Sensitivity Air Sensitive
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material Indole-7-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.