missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-carbonitrile, 98+%

Product Code. 11354536
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354536 5 g 5g
11344536 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354536 Supplier Thermo Scientific Alfa Aesar Supplier No. L20099.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 15861-36-6 | C9H6N2 | 142.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15861-36-6
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00016732
InChI Key SZSZDBFJCQKTRG-UHFFFAOYSA-N
Synonym 6-cyanoindole, 6-indolecarbonitrile, indole-6-carbonitrile, 6-cyano-1h-indole, indole-6-cyano, 6-cyano-indole, 6-indole carbonitrile, pubchem7239, cyanoindole-6, 1h-indol-6-yl cyanide
PubChem CID 85146
IUPAC Name 1H-indole-6-carbonitrile
SMILES C1=CC(=CC2=C1C=CN2)C#N

Specifications

Melting Point 128°C to 132°C
Odor Odorless
Quantity 5 g
Beilstein 116502
Sensitivity Light sensitive
Formula Weight 142.16
Percent Purity ≥98%
Chemical Name or Material Indole-6-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.