missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-boronic acid, 98%

Product Code. p-7035594
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480058 250 mg 250mg
15470058 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480058 Supplier Thermo Scientific Alfa Aesar Supplier No. H53313.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 147621-18-9 | C8H8BNO2 | 160.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147621-18-9
Molecular Formula C8H8BNO2
Molecular Weight (g/mol) 160.97
MDL Number MFCD03095176
InChI Key ZVMHOIWRCCZGPZ-UHFFFAOYSA-N
Synonym indole-6-boronic acid, 6-indolylboronic acid, 1h-indol-6-yl boronic acid, 6-indoleboronic acid, indole-6-boronicacid, 1h-indol-6-yl-6-boronic acid, 6-borono-1h-indole, 6-indole boronic acid, 6-boronoindole, 1h-indole-6-boronic acid
PubChem CID 2763205
IUPAC Name 1H-indol-6-ylboronic acid
SMILES OB(O)C1=CC=C2C=CNC2=C1

Specifications

Melting Point 177°C to 181°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 160.97
Percent Purity 98%
Chemical Name or Material Indole-6-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.