missing translation for 'onlineSavingsMsg'
Learn More

Indole-5-carboxylic acid, 98%

Product Code. 11498567
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498567 25 g 25g
11478567 1 g 1g
11488567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498567 Supplier Thermo Scientific Alfa Aesar Supplier No. A18627.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1670-81-1 | C9H6NO2 | 160.15 g/mol

Indole-5-carboxylic acid is the suitable reagent used to study the intermolecular excited state proton transfer in indole-2-carboxylic acid and indole-5-carboxylic acid in various solvents in acidic, basic, and neutral media by steady state and time resolved fluorescence spectroscopy. It may be used in the electrochemical synthesis of poly(indole-5-carboxylic acid) (PICA) films. Also used as reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, as potential anticancer immunomodulators, synthesis of indirubin derivatives and amide conjugates with ketoprofen, as inhibitors of Gli1-mediated transcription in Hedgehog pathway.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-5-carboxylic acid is the suitable reagent used to study the intermolecular excited state proton transfer in indole-2-carboxylic acid and indole-5-carboxylic acid in various solvents in acidic, basic, and neutral media by steady state and time resolved fluorescence spectroscopy. It may be used in the electrochemical synthesis of poly(indole-5-carboxylic acid) (PICA) films. Also used as reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, as potential anticancer immunomodulators, synthesis of indirubin derivatives and amide conjugates with ketoprofen, as inhibitors of Gli1-mediated transcription in Hedgehog pathway.

Solubility
Soluble in ethanol (50 mg/ml), dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1670-81-1
Molecular Formula C9H6NO2
Molecular Weight (g/mol) 160.15
MDL Number MFCD00005678
InChI Key IENZCGNHSIMFJE-UHFFFAOYSA-M
Synonym indole-5-carboxylic acid, 5-carboxyindole, 5-indolecarboxylic acid, indole-5-carboxylic aicd, pubchem1694, 5-carboxy-1h-indole, indole-5-carboxylicacid, indol-5-carboxylic acid, 5-indole carboxylic acid, indole 5-carboxylic acid
PubChem CID 74280
IUPAC Name 1H-indole-5-carboxylic acid
SMILES [O-]C(=O)C1=CC=C2NC=CC2=C1

Specifications

Melting Point 208°C to 212°C
Quantity 25 g
Beilstein 124391
Solubility Information Soluble in ethanol (50mg/ml),dimethyl sulfoxide and methanol.
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material Indole-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.