missing translation for 'onlineSavingsMsg'
Learn More

Indole-5-carbonitrile, 98+%

Product Code. 11454837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454837 5 g 5g
11444837 1 g 1g
11464837 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454837 Supplier Thermo Scientific Alfa Aesar Supplier No. B21810.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15861-24-2 | C9H6N2 | 142.161 g/mol

Reactant used for parallel synthesis of dihydroisoquinolines via silver and L-proline co-catalyzed three-component coupling reaction, preparation of novel PPARα/γ dual agonists for potential treatment of metabolic syndrome and IDDM, chemo selective and regioselective preparation of benzoyl indoles, 4,5-dihydrocyclopenta[c]quinolines by palladium-catalyzed ring-expansion reaction alkynes, using O2 as the oxidant and vinylindoles by hydroarylation of alkynes using indium bromide catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for parallel synthesis of dihydroisoquinolines via silver and L-proline co-catalyzed three-component coupling reaction, preparation of novel PPARα/γ dual agonists for potential treatment of metabolic syndrome and IDDM, chemo selective and regioselective preparation of benzoyl indoles, 4,5-dihydrocyclopenta[c]quinolines by palladium-catalyzed ring-expansion reaction alkynes, using O2 as the oxidant and vinylindoles by hydroarylation of alkynes using indium bromide catalyst.

Solubility
Soluble in chloroform, hexane and methanol. Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15861-24-2
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00005669
InChI Key YHYLDEVWYOFIJK-UHFFFAOYSA-N
Synonym 5-cyanoindole, indole-5-carbonitrile, 5-cyano indole, 5-cyano-1h-indole, indole-5-cyano, 5-indolecarbonitrile, 5-cyano-indole, indol-5-carbonitrile, indole, 5-cyano, pubchem7326
PubChem CID 27513
IUPAC Name 1H-indole-5-carbonitrile
SMILES C1=CC2=C(C=CN2)C=C1C#N

Specifications

Melting Point 104°C to 108°C
Odor Odorless
Quantity 5 g
Beilstein 116738
Sensitivity Light sensitive
Solubility Information Soluble in chloroform,hexane and methanol. Insoluble in water.
Formula Weight 142.16
Percent Purity ≥98%
Chemical Name or Material Indole-5-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.