missing translation for 'onlineSavingsMsg'
Learn More

Indole-5-boronic acid, 95%

Product Code. 15453587
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453587 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15453587 Supplier Thermo Scientific Alfa Aesar Supplier No. H27008.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 144104-59-6 | C8H8BNO2 | 160.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 144104-59-6
Melting Point 170°C to 175°C
Molecular Formula C8H8BNO2
MDL Number MFCD01319013
Quantity 1 g
Sensitivity Air sensitive
Synonym 5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid
InChI Key VHADYSUJZAPXOW-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C2NC=CC2=C1
IUPAC Name 1H-indol-5-ylboronic acid
Molecular Weight (g/mol) 160.97
PubChem CID 2734361
Formula Weight 160.97
Percent Purity 95%
Chemical Name or Material Indole-5-boronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.