missing translation for 'onlineSavingsMsg'
Learn More

Indole-4-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10163194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163194 1 g Glass bottle 1g
10264814 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163194 Supplier Thermo Scientific Acros Supplier No. 398060010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2124-55-2
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00009739
InChI Key ROGHUJUFCRFUSO-UHFFFAOYSA-N
Synonym indole-4-carboxylic acid, indole-4-carboxylicacid, 4-carboxyindole, 1h-4-indolecarboxylic acid, 1h-indole-4-carboxylic?acid, 4-indolecarboxylic acid, pubchem1693, acmc-209fio, 4-indole-carboxylic acid, indol-4-yl carboxylic acid
PubChem CID 595229
IUPAC Name 1H-indole-4-carboxylic acid
SMILES C1=CC(=C2C=CNC2=C1)C(=O)O

Specifications

Melting Point 207°C to 209°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Indole-4-carboxylic acid
compliance-icons
Product Identifier
  • Indole-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.