missing translation for 'onlineSavingsMsg'
Learn More

Indole-4,5,6,7-d4, TRC

Product Code. 30392653
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392653 5 mg 5mg
30394771 10 mg 10mg
30395581 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392653 Supplier Toronto Research Chemicals Supplier No. TRCI5773215MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73509-22-5
Molecular Formula C8H3D4N
Molecular Weight (g/mol) 121.17
Synonym 1H-Indole-d4, 1-Azaindene-d4, 1-Benzazole-d4, 2,3-Benzopyrrole-d4, Benzo[b]pyrrole-d4, Ketole-d4, NSC 1964-d4
SMILES [2H]C1=C2C(NC=C2)=C([2H])C([2H])=C1[2H]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H/i1D,2D,3D,4D
Chemical Name or Material Indole-4,5,6,7-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.