missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-3-pyruvic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Indole-3-pyruvic Acid |
| CAS | 392-12-1 |
| Molecular Formula | C11 H9 N O3 |
| Quantity | 100 mg |
| Synonym | α-Oxo-1H-indole-3-propanoic Acid, Indolepyruvic Acid, 1H-Indole-3-pyruvic Acid, 3-(3-Indolyl)-2-oxopropanoic Acid, 3-(3-Indolyl)pyruvic Acid, 3-Indolylpyroracemic Acid, 3-Indolylpyruvic Acid, Indole-3-pyruvate, Indolyl-3-pyruvate, NSC 88874, POL 240, β-Indolepyruvic Acid, β-Indolylpyruvic Acid |
| InChI Formula | InChI=1S/C11H9NO3/c13-10(11(14)15)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,12H,5H2,(H,14,15) |
| SMILES | OC(=O)C(=O)Cc1c[nH]c2ccccc12 |
| IUPAC Name | 3-(1H-indol-3-yl)-2-oxo-propanoic acid |
| Molecular Weight (g/mol) | 203.19 |
| Formula Weight | 203.0582 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?