missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxylic Acid, TRC

Product Code. 30391143
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391143 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391143 Supplier Toronto Research Chemicals Supplier No. TRCI62900025G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Indole-3-carboxylic Acid
CAS 771-50-6
Molecular Formula C9 H7 N O2
Quantity 25 g
Synonym 1H-Indole-3-carboxylic acid, Tropisetron Hydrochloride Imp. B (EP), Indole-3-carboxylic acid (8CI), 3-Carboxyindole, 3-Indoleformic acid, 3-Indolylcarboxylic acid, Indol-3-carboxylic acid, Indole-β-carboxylic acid, β-Indolylcarboxylic acid
InChI Formula InChI=1S/C9H7NO2/c11-9(12)7-5-10-8-4-2-1-3-6(7)8/h1-5,10H,(H,11,12)
SMILES OC(=O)c1c[nH]c2ccccc12
IUPAC Name 1H-indole-3-carboxylic acid
Molecular Weight (g/mol) 161.16
Formula Weight 161.0477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.