missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxylic acid, 98%

Product Code. 11470473
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470473 25 g 25g
11450473 1 g 1g
11460473 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470473 Supplier Thermo Scientific Alfa Aesar Supplier No. A12634.14
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 771-50-6 | C9H7NO2 | 161.16 g/mol

Indole-3-carboxylic acid is used as a raw material in synthesis. The structures of the derivatives of indole-3-carboxylic acid were studied using gas chromatography with high resolution mass spectrometry (GC-HRMS), ultra-high performance liquid chromatography in combination with high resolution tandem mass spectrometry (UHPLC-HRMS), nuclear magnetic resonance spectroscopy (NMR) and Fourier transform infrared spectroscopy. Reactant for preparation of: Anticancer agents, Derivatives of amino acids and peptides, Serotonin 5-HT4 receptor antagonists, Primary acylureas, Inhibitors of Gli1-mediated transcription in the Hedgehog pathway, Serotonin 5-HT6 antagonists, Very Late Antigen-4 (VLA-4) sntagonists, EphB3 receptor tyrosine kinase inhibitors, Potential Therapeutic Agent for Alzheimer?s Disease, Vinyl ester pseudotripeptide proteasome inhibitors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-carboxylic acid is used as a raw material in synthesis. The structures of the derivatives of indole-3-carboxylic acid were studied using gas chromatography with high resolution mass spectrometry (GC-HRMS), ultra-high performance liquid chromatography in combination with high resolution tandem mass spectrometry (UHPLC-HRMS), nuclear magnetic resonance spectroscopy (NMR) and Fourier transform infrared spectroscopy. Reactant for preparation of: Anticancer agents, Derivatives of amino acids and peptides, Serotonin 5-HT4 receptor antagonists, Primary acylureas, Inhibitors of Gli1-mediated transcription in the Hedgehog pathway, Serotonin 5-HT6 antagonists, Very Late Antigen-4 (VLA-4) sntagonists, EphB3 receptor tyrosine kinase inhibitors, Potential Therapeutic Agent for Alzheimer′s Disease, Vinyl ester pseudotripeptide proteasome inhibitors

Solubility
Soluble in 95% Ethanol: 50 mg/ml, also soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-50-6
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005624
InChI Key KMAKOBLIOCQGJP-UHFFFAOYSA-N
Synonym indole-3-carboxylic acid, 3-indoleformic acid, 3-carboxyindole, indole-3-carboxylicacid, 3-indolecarboxylic acid, indole-3-carboxylate, 3-indolylcarboxylic acid, beta-indolylcarboxylic acid, indole-3 carboxylic acid, ico
PubChem CID 69867
ChEBI CHEBI:24809
IUPAC Name 1H-indole-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CN2)C(=O)O

Specifications

Melting Point ∼239°C (decomposition)
Quantity 25 g
Beilstein 129435
Solubility Information Soluble in 95% Ethanol: 50mg/ml,also soluble in methanol.
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material Indole-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.