missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxamidoxime, 97%, Thermo Scientific™

Product Code. 15480728
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480728 250 mg 250mg
15470728 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480728 Supplier Thermo Scientific Alfa Aesar Supplier No. H54429.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95649-37-9
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.191
MDL Number MFCD00460181
InChI Key DRCFXQBNDIRJCT-CLFYSBASSA-N
Synonym n'-hydroxy-1h-indole-3-carboximidamide, n-amino indol-3-ylidene methyl hydroxylamine, n-hydroxy-1h-indole-3-carboximidamide, 3-indolecarboxylic acid amidoxime, n-hydroxy-1h-indole-3-carboxamidine, z-n'-hydroxy-1h-indole-3-carboximidamide
PubChem CID 6097083
IUPAC Name N-[(E)-amino(indol-3-ylidene)methyl]hydroxylamine
SMILES C1=CC=C2C(=C1)C(=C(N)NO)C=N2

Specifications

Quantity 250 mg
Beilstein 4249031
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material Indole-3-carboxamidoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.