missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldehyde, TRC

Product Code. 30137247
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137247 10 g 10g
30132530 1 g 1g
30009268 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137247 Supplier Toronto Research Chemicals Supplier No. TRCI61499010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-89-8
Molecular Formula C9 H7 N O
Molecular Weight (g/mol) 145.16
Synonym 1H-Indole-3-carboxaldehyde, Indole-3-carboxaldehyde (6CI,8CI), 1H-Indol-3-yl carboxaldehyde, 1H-Indole-3-aldehyde, 1H-Indole-3-carbaldehyde, 1H-Indole-3-methanal, 3-Formyl-1H-indole, 3-Formylindole, 3-Indolylformaldehyde, Indole-3-aldehyde, Indole-3-carbaldehyde, Indole-3-carboxylaldehyde, Indole-3-formaldehyde, NSC 10118, β-Indolylaldehyde
IUPAC Name 1H-indole-3-carbaldehyde
SMILES O=Cc1c[nH]c2ccccc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H7NO/c11-6-7-5-10-9-4-2-1-3-8(7)9/h1-6,10H
Formula Weight 145.0528
Percent Purity >95%
Chemical Name or Material Indole-3-carboxaldehyde
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.