missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldehyde, 97%

Product Code. 10553131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553131 25 g Plastic bottle 25g
10349343 5 g Glass bottle 5g
10512941 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10553131 Supplier Thermo Scientific Acros Supplier No. 156640250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 487-89-8 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-89-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00005622
InChI Key OLNJUISKUQQNIM-UHFFFAOYSA-N
Synonym indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
PubChem CID 10256
ChEBI CHEBI:28238
IUPAC Name 1H-indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=CN2)C=O

Specifications

Melting Point 193°C to 199°C
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 21, 313
Solubility Information Solubility in water: insoluble
Formula Weight 145.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Indole-3-carboxaldehyde, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.