missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carbonitrile, 98%

Product Code. 11418696
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418696 5 g 5g
11408696 1 g 1g
11428696 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418696 Supplier Thermo Scientific Alfa Aesar Supplier No. A16041.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5457-28-3 | C9H6N2 | 142.16 g/mol

Indole-3-carbonitrile is used as a synthesis reagent for preparation of 4-substituted β-lactams, biologically active indoles, inhibitors of glycogen synthase kinase 3β (GSK-3), indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors and HIV-1 integrase inhibitors. It is also used as a reactant for intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques and for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-carbonitrile is used as a synthesis reagent for preparation of 4-substituted β-lactams, biologically active indoles, inhibitors of glycogen synthase kinase 3β (GSK-3), indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors and HIV-1 integrase inhibitors. It is also used as a reactant for intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques and for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5457-28-3
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00022717
InChI Key CHIFTAQVXHNVRW-UHFFFAOYSA-N
Synonym 3-cyanoindole, indole-3-carbonitrile, 3-indolecarbonitrile, chembl46724, zlchem 360, 3-cyano-1h-indole, pubchem7325, acmc-209lhi, 1h-indol-3-yl cyanide
PubChem CID 230282
IUPAC Name 1H-indole-3-carbonitrile
SMILES N#CC1=CNC2=CC=CC=C12

Specifications

Melting Point 177°C to 182°C
Odor Odorless
Quantity 5 g
Beilstein 120888
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 142.16
Percent Purity 98%
Chemical Name or Material Indole-3-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.