Learn More
Indole-3-carbonitrile, 98%
Description
Indole-3-carbonitrile is used as a synthesis reagent for preparation of 4-substituted β-lactams, biologically active indoles, inhibitors of glycogen synthase kinase 3β (GSK-3), indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors and HIV-1 integrase inhibitors. It is also used as a reactant for intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques and for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indole-3-carbonitrile is used as a synthesis reagent for preparation of 4-substituted β-lactams, biologically active indoles, inhibitors of glycogen synthase kinase 3β (GSK-3), indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors and HIV-1 integrase inhibitors. It is also used as a reactant for intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques and for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications
Specifications
| Melting Point | 177°C to 182°C |
| Odor | Odorless |
| Quantity | 1 g |
| Beilstein | 120888 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 142.16 |
| Percent Purity | 98% |
| Chemical Name or Material | Indole-3-carbonitrile |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.