missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carbinol, MedChemExpress

Product Code. 16452509
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452509 1 g 1g
16451496 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452509 Supplier Medchem Express Supplier No. HYN0170
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Indole-3-carbinol (I3C) inhibits NF-κB activity and also is an Aryl hydrocarbon receptor (AhR) agonist, and an inhibitor of WWP1 (WW domain-containing ubiquitin E3 ligase 1).

Indole-3-carbinol (I3C) inhibits NF-κB activity and also is an Aryl hydrocarbon receptor (AhR) agonist, and an inhibitor of WWP1 (WW domain-containing ubiquitin E3 ligase 1).

Specifications

Chemical Name or Material Indole-3-carbinol
CAS 700-06-1
Color White
Purity Grade Notes Research
Molecular Formula C9H9NO
Quantity 1 g
Synonym I3C 3-Indolemethanol
Solubility Information DMSO : 100 mg/mL (679.49 mM; Need ultrasonic)
SMILES OCC1=CNC2=C1C=CC=C2
Molecular Weight (g/mol) 147.17
Formula Weight 147.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.