missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-butyric Acid, TRC

Product Code. 30041215
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041215 10 mg 10mg
30003561 1 g 1g
30010718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041215 Supplier Toronto Research Chemicals Supplier No. TRCI57780010MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-32-4
Molecular Formula C12 H13 N O2
Molecular Weight (g/mol) 203.24
IUPAC Name 4-(1H-indol-3-yl)butanoic acid
SMILES OC(=O)CCCc1c[nH]c2ccccc12

Specifications

Quantity 10 mg
Synonym 1H-Indole-3-butanoic acid (9CI, ACI), Indole-3-butyric acid (8CI), 1H-Indole-3-butyric acid, 3-Indolyl-γ-butyric acid, 3-Indolylbutyric acid, 4-(1H-Indol-3-yl)butanoic acid, 4-(1H-Indol-3-yl)butyric acid, 4-(3-Indolyl)butanoic acid, 4-(Indol-3-yl)butyric acid, [3-(3-Indolyl)propyl]carboxylic acid, Clonex, Clonex (rooting hormone), Hormex, Hormodin, IBA, Indole-3-butanoic acid, Indolebutyric acid, Kornevin, NSC 3130, Oxyberon, Rootex, Seradix, Seradix 2, Seradix 3, Seradix B 2, Seradix B 3, Stim-Root, β-IBA, β-Indolebutyric acid, β-Indolylbutyric acid, γ-(Indol-3-yl)butyric acid, γ-(Indole-3)-butyric acid
InChI Formula InChI=1S/C12H13NO2/c14-12(15)7-3-4-9-8-13-11-6-2-1-5-10(9)11/h1-2,5-6,8,13H,3-4,7H2,(H,14,15)
Formula Weight 203.0946
Chemical Name or Material 4-(3-Indolyl)-butyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.