missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-butyric Acid Potassium Salt, TRC

Product Code. 30395929
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395929 5 g -
30405891 1 g -
30394329 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395929 Supplier Toronto Research Chemicals Supplier No. TRCI5777655G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60096-23-3
Molecular Formula C12H12KNO2
Molecular Weight (g/mol) 241.33
Synonym 1H-Indole-3-butanoic Acid Potassium Salt, 1H-Indole-3-butanoic Acid Monopotassium Salt, 3-Indolebutyric Acid Potassium Salt, IBA K Salt, IBA Potassium Salt
IUPAC Name potassium;4-(1H-indol-3-yl)butanoate
SMILES [K+].[O-]C(=O)CCCc1c[nH]c2ccccc12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H13NO2.K/c14-12(15)7-3-4-9-8-13-11-6-2-1-5-10(9)11;/h1-2,5-6,8,13H,3-4,7H2,(H,14,15);/q;+1/p-1
Formula Weight 241.0505
Chemical Name or Material Indole-3-butyric Acid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.