missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-acetyl Beta-D-Glucopyranose, TRC

Product Code. 30009687
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009687 2.5 mg 2.5mg
30004241 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009687 Supplier Toronto Research Chemicals Supplier No. TRCI6270202.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 19817-95-9
Molecular Formula C16 H19 N O7
Molecular Weight (g/mol) 337.32
Synonym 1-(1H-Indole-3-acetate) β-D-Glucopyranose, 1-O-(Indol-3'-ylacetyl) -β-D-glucopyranose, Indole-3-acetic Acid β-D-Glucopyranosyl Ester, Indole-3-acetic Acid β-Glucoside;
IUPAC Name [(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl] 2-(1H-indol-3-yl)acetate
SMILES C(C(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)=O)C=2C=3C(NC2)=CC=CC3

Specifikationer

Quantity 2.5 mg
InChI Formula InChI=1S/C16H19NO7/c18-7-11-13(20)14(21)15(22)16(23-11)24-12(19)5-8-6-17-10-4-2-1-3-9(8)10/h1-4,6,11,13-18,20-22H,5,7H2/t11-,13-,14+,15-,16+/m1/s1
Formula Weight 337.1162
Percent Purity >95%
Chemical Name or Material Indole-3-acetyl beta-D-Glucopyranose
Grade HPLC
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.